Would you like to log in again?
Immunotag™ Mouse ANA (Anti-nuclear Anti-body) ELISA Kit, G-Biosciences
Supplier: G-Biosciences
Immunotag™
IT6147
76259-102EA 776.86 USD InStock
76259-102
Immunotag™ Mouse ANA (Anti-nuclear Anti-body) ELISA Kit, G-Biosciences
Assays ELISAs
  • Detection method: Sandwich ELISA, double antigen
  • Range: 0.469 to 30 ng/ml
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty