Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead(II) nitrate ≥99%, Puriss., Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Lead dinitrate

11520-1KG
BJ11520-1KGEA 163.24 USD InStock
BJ11520-1KG
Lead(II) nitrate ≥99%, Puriss., Fluka™
Lead (II) nitrate


Formula: Pb(NO₃)₂
MW: 331.21 g/mol
Boiling Pt: Decomposes on heating.
Melting Pt: 458-459 °C (1.023 hPa)
Density: 4.49 g/cm³ (20.1 °C)
Storage Temperature: Room Temperature
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1,II
Order Now
Specification Test Results
Assay Min. 99 %
Iron (Fe) Max. 0.002 %
Chloride (Cl) Max. 0.002 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty