Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disialoganglioside GD2 (human brain) ≥98% (by TLC), Calbiochem®

Supplier: MilliporeSigma
Synonyms: GalNAcβ1,4(Neu5Acα2,8)(Neu5Acα2,03)Galβ1,4Glcβ1,1ʹ CeramideGanglioside GD₂, Disialo

345743-500UG
80502-554EA 802.33 USD InStock
80502-554
Disialoganglioside GD2 (human brain) ≥98% (by TLC), Calbiochem®
Disialoganglioside GD2
Gangliosides are highly purified sialic acid-containing glycolipids that are useful as markers of various cell types and antigens. Prepared from tissue of individuals that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Used as markers of various cell types and antigens.


Formula: C₇₈H₁₃₈N₄O₃₄
Storage Temperature: Freezer
CAS Number: 65988-71-8
Order Now
Specification Test Results
Purity (TLC) ≥98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty