Would you like to log in again?
Thermometer Clamp, United Scientific Supplies
Supplier: United Scientific Supplies
THCL01-VWR
76293-412EA 11.39 USD InStock
76293-412
Thermometer Clamp, United Scientific Supplies
Clamps General Purpose Clamps
Support a wide range of thermometers.
  • Jaws rotate 360 degrees
  • Holds thermometers gently without breaking
Thermometer Clamp features adjustable 360° rotating jaw for suspending thermometers. The clamp attaches to rods up to 13 mm (1/2") O.D.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty