Would you like to log in again?
Borrelia burgdorferi Recombinant p39 Control Protein (from E.coli)
Supplier: Rockland Immunochemicals
000-001-C17
RL000-001-C17EA 444 USD InStock
RL000-001-C17
Borrelia burgdorferi Recombinant p39 Control Protein (from E.coli)
Proteins and Peptides
The p39 protein, or Basic membrane protein A, is one of the immunogenic cell membrane components of Borrelia burgdorferi, the spirochete carried by Ixodes ticks. The spirochete migrates from the tick midgut during feeding to its salivary glands and are thus transmitted to the mammal host. This transition may be facilitated by changes in expression of some B. burgdorferi genes. It is believed that expression of the various proteins associated with the spirochete may be regulated by the changes in tick life cycle, changes in conditions during tick feeding (such as temperature, pH, and nutrients) and/or in coordination with the course of infection of the mammal host. BmpA is expressed during the invasion of the spirochete and in the evolution of the arthritis of Lyme disease in mammals. It belongs to the BMP lipoprotein family. The major products of the B. burgdorferi basic membrane protein (bmp) A/B operon that are induced in murine and human joints possess inflammatory properties. Non-lipidated and lipidated versions of BmpA have been shown to induce the pro-inflammatory cytokine TNF-α and IL-1ß in human synovial cells. The induction of cytokine responses in synovial cells via activation of the NF-kappaB and p38 MAP kinase pathways could potentially contribute to the genesis of Lyme arthritis. The BmpA outer-surface protein is an important antigen for serodiagnosis of human infection. B. burgdorferi adheres to host extracellular matrix components, including laminin, but may not bind mammalian type I or type IV collagens or fibronectin. Lyme disease proteins are ideal for researchers interested in immunology, neurology, and rheumatology, coinfections , autoimmune, and neurodegenerative diseases.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty