Would you like to log in again?
Borrelia burgdorferi Recombinant Surface Lipoprotein p27 Control Protein (from E.coli)
Supplier: Rockland Immunochemicals
000-001-C30
RL000-001-C30EA 444 USD InStock
RL000-001-C30
Borrelia burgdorferi Recombinant Surface Lipoprotein p27 Control Protein (from E.coli)
Proteins and Peptides
Surface Lipoprotein p27 of Borrelia burgdorferi is a surface-exposed lipoprotein that has been shown (by Western blot and Northern blot) to be expressed in the European B. burgdorferi strain B29, but not in the American strain B31. Cell envelope proteins of bacterial pathogens play important roles in the host-parasite interactions that occur during infection, including cell adherence, cell invasion, and immune cell activation or evasion. p27 is a basic protein of 248 amino acids with a typical prokaryotic leader sequence of 17 amino acid residues at the N-terminus of the proposed translation product. The p27 gene is located on a linear plasmid of a size of approximately 55 kb. Borrelia spirochetes are unique among diderm bacteria in their abundance of surface-displayed lipoproteins, some of which play important roles in the pathogenesis of Lyme disease and relapsing fever. There is evidence that Borrelia lipoproteins are specifically targeted to the bacterial surface, but that they can be retained in the periplasm by sequence-specific signals. Lyme disease proteins are ideal for researchers interested in immunology, neurology, and rheumatology, coinfections , autoimmune, and neurodegenerative diseases.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty