Would you like to log in again?
Human Recombinant AKT3 (from Baculovirus (Insect cells))
Supplier: Rockland Immunochemicals
009-001-P24
10799-950EA 362 USD InStock
10799-950
Human Recombinant AKT3 (from Baculovirus (Insect cells))
Proteins and Peptides
AKT3 is a component of the PI-3 kinase pathway and is activated by phosphorylation at Ser 473 and Thr 308. AKT is a cytoplasmic protein also known as Protein Kinase B (PKB) and RAC (Related to A and C kinases). AKT is a key regulator of many signal transduction pathways, and it exhibits tight control over cell proliferation and cell viability. Overexpression or inappropriate activation of AKT is noted in many types of cancer. AKT mediates many of the downstream events of PI 3-kinase (a lipid kinase activated by growth factors, cytokines and insulin). PI 3-kinase recruits AKT to the membrane, where it is activated by PDK1 phosphorylation. Once phosphorylated, AKT dissociates from the membrane and phosphorylates targets in the cytoplasm and the cell nucleus. AKT has two main roles: (i) inhibition of apoptosis; (ii) promotion of proliferation. AKT3 recombinant protein is ideal for investigators involved in Cell Signaling, Neuroscience and Signal Transduction research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty