Would you like to log in again?
Human Recombinant YWHAB (from E. coli)
Supplier: Rockland Immunochemicals
009-001-R24 009-001-R24S
10799-446EA 313 USD InStock
10799-446 10799-448
Human Recombinant YWHAB (from E. coli)
Proteins and Peptides
14-3-3α/β is a member of the highly conserved 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins (1). 14-3-3α/β protein has been shown to interact with RAF1 and CDC25 phosphatases, suggesting that it may play a role in linking mitogenic signaling and the cell cycle machinery. 14-3-3-β interacts with the TSC1-TSC2 dimer (2). The interaction required phosphorylation of TSC2 at Ser1210. Binding of 14-3-3-β to TSC2 did not alter the interaction between TSC1 and TSC2, but it reduced the ability of the complex to inhibit phosphorylation of ribosomal protein S6 kinase impairing the ability of the complex to inhibit cell growth. 14-3-3 alpha/beta Protein is ideal for investigators involved in Cell Stress & Chaperone Proteins, Cell Signaling, Cancer research, Cell Cycle, Cellular Stress, Neurobiology, and WNT Signaling research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty