Would you like to log in again?
Human Recombinant YWHAE (from E. coli)
Supplier: Rockland Immunochemicals
009-001-R26 009-001-R26S
10799-456EA 513 USD InStock
10799-456 10799-454
Human Recombinant YWHAE (from E. coli)
Proteins and Peptides
14-3-3ε also known as tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein is a member of the 14-3-3 family. 14-3-3ε has been reported to be involved in heat shock transcription factor 1 (HSF1) and extracellular signal regulated protein kinase (ERK) pathways (1). 14-3-3ε interaction with the deubiquitinating enzyme UBPY, also known as USP8, regulates cargo sorting and membrane traffic at early endosomes. Association of 14-3-3ε, HSF1 with ERK during heat shock may modulate the amplitude of the response and control the termination of HSP expression on resumption of growth conditions (2). 14-3-3ε protein is ideal for investigators involved in Cell Stress & Chaperone Proteins, Cell Signaling, Cancer research, Cell Cycle, Cellular Stress, Neurobiology, and WNT Signaling research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty