Would you like to log in again?
Human Recombinant YWHAQ (from E. coli)
Supplier: Rockland Immunochemicals
009-001-R29S 009-001-R29
10799-474EA 313 USD InStock
10799-474 10799-476
Human Recombinant YWHAQ (from E. coli)
Proteins and Peptides
14-3-3θ (also known as tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide) is a member of the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. Through interaction with ASK1, c-jun NH-terminal kinase, and p38 mitogen-activated protein kinase (MAPK), 14-3-3θ plays an important role in controlling apotopsis (1). Induced expression of 14-3-3θ protein has been reported in patients with amyotrophic lateral sclerosis. Additionally, 14-3-3θ has been observed to mediate nucleocytoplasmic shuttling of the N protein (coronavirus nucleocapsid protein) which causes severe acute respiratory syndrome (2). 14-3-3θ Protein is ideal for investigators involved in Cell Stress& Chaperone Proteins, AKT/PKB Pathway, Cancer, Cell Cycle, Cellular Stress, ERK/MAPK Pathway, Neurobiology, PKA/PKC Pathway, and WNT Signaling research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty