Would you like to log in again?
Human Recombinant BAX (from E. coli)
Supplier: Rockland Immunochemicals
009-001-R47 009-001-R47S
10802-020EA 714 USD InStock
10802-020 10802-018
Human Recombinant BAX (from E. coli)
Proteins and Peptides
BAX is a proapoptotic protein of the BCL2 protein family. BAX forms a heterodimer with BCL2 and functions as an apoptotic activator. BAX interacts with and increases the opening of the mitochondrial voltage-dependent anion channel (VDAC), which leads to the loss in the mitochondrial membrane potential and the release of cytochrome c (1). The expression of BAX gene is regulated by the tumor suppressor p53 and BAX has been shown to be involved in p53-mediated apoptosis. Multiple alternatively spliced transcript variants, which encode different isoforms, have been reported for this gene (2). BAX Protein is ideal for investigators involved in Signaling Proteins, Apoptosis Proteins, AKT/PKB Pathway, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, and Neurobiology research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty