Would you like to log in again?
Human Recombinant CALM (from E. coli)
Supplier: Rockland Immunochemicals
009-001-R55 009-001-R55S
10800-506EA 313 USD InStock
10800-506 10800-508
Human Recombinant CALM (from E. coli)
Proteins and Peptides
Calmodulin 1 is a member of calcium-modulated proteins which is present in the cytosol and on membranes facing the cytosol and has a high affinity for calcium. Calmodulin 1 has 4 calcium-binding domains and plays a role in cell growth, cell cycle, signal transduction and the synthesis and release of neurotransmitters. Calmodulin can bind to the epidermal growth factor receptor at its cytosolic juxtamembrane region and this inhibits its tyrosine kinase activity (1). A number of other proteins including HSP70 have been shown to interact with Calmodulin 1 in a cell-phase-specific manner (2). Calmodulin 1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cardiovascular Disease, ERK/MAPK Pathway, Neurobiology, and PKA/PKC Pathway research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty