Would you like to log in again?
Human Recombinant CDKL1 (from Baculovirus (Sf9 Insect Cells))
Supplier: Rockland Immunochemicals
009-001-R66S 009-001-R66
10800-730EA 364 USD InStock
10800-730 10800-732
Human Recombinant CDKL1 (from Baculovirus (Sf9 Insect Cells))
Proteins and Peptides
CDKL1 or cyclin-dependent kinase-like 1 is a member of a large family of CDC2-related serine/threonine protein kinases. CDKL1, which is also known as p42 KKIALRE, accumulates primarily in the nucleus and is known to regulate important transitions in the eukaryotic cell cycle (1). CDKL1 and the related CDC2 protein kinase have a similar pattern of mRNA expression except in the reproductive organs. CDKL1 is expressed in several human tissues, with the highest expression in kidney and ovary (2). CDKL1 is overexpressed in breast cancer patients and has a positive detection efficiency of 77%. Inhibiting CDKL1 function with shRNA in MCF-7 cells shows increased accumulation of the cells at the G2/M phase and increased sensitivity to cell cycle chemotherapeutic drugs. CDKL1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, Cell Cycle, and Ser/Thr Kinases research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty