Would you like to log in again?
Human Recombinant CDKL3 (from Baculovirus (Sf9 Insect Cells))
Supplier: Rockland Immunochemicals
009-001-R68S 009-001-R68
10800-738EA 364 USD InStock
10800-738 10800-740
Human Recombinant CDKL3 (from Baculovirus (Sf9 Insect Cells))
Proteins and Peptides
CDKL3 or cyclin-dependent kinase-like 3 is a member of cyclin-dependent protein kinase (CDK) family which is highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and is known to be important regulators of cell cycle progression and cellular proliferation. CDKL3 is a novel CDC2-related kinase with features of both mitogen-activated protein kinases and cyclin-dependent kinases (1). CDKL3 is identified as a gene absent in leukemic patients with chromosome 5q deletion and this loss may be an important determinant of dysmyelopoiesis (2). CDKL3 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, Cell Cycle, and Ser/Thr Kinases research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty