Would you like to log in again?
Human Recombinant CDKN3 (from Baculovirus (Sf9 Insect Cells))
Supplier: Rockland Immunochemicals
009-001-R70 009-001-R70S
10800-752EA 364 USD InStock
10800-752 10800-754
Human Recombinant CDKN3 (from Baculovirus (Sf9 Insect Cells))
Proteins and Peptides
CDKN3 is a member of the dual specificity protein phosphatase family that acts as a cyclin-dependent kinase inhibitor (1). CDKN3 has been shown to interact with and dephosphorylate specifically the CDK2 kinase thereby preventing it’s activation. CDKN3 has been reported to be deleted, mutated, or overexpressed in several kinds of cancers. Increased expression of CDKN3 leads to increased levels of kinase-associated phosphatase activity that inhibits the G(1)/S transition of the cell cycle by dephosphorylating the cyclin-dependent kinases (2). CDKN3 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, Cell Cycle, and Ser/Thr Kinases research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty