Would you like to log in again?
Human Recombinant CFL1 (from E. coli)
Supplier: Rockland Immunochemicals
009-001-R72S 009-001-R72
10802-082EA 364 USD InStock
10802-082 10802-084
Human Recombinant CFL1 (from E. coli)
Proteins and Peptides
Cofilin 1 (non-muscle), also known as CFL1, is a widely distributed intracellular actin-modulating protein that binds and depolymerizes filamentous F-actin and inhibits the polymerization of monomeric G-actin in a pH-dependent manner (1). It is involved in the translocation of actin-cofilin complex from cytoplasm to nucleus (2). CFL1 processed proteins are most probably produced by HIV-1 protease digestion. Cofilin 1 Protein is ideal for investigators involved in Signaling Proteins, Microtubule/Actin Associated Proteins, Cancer, Cardiovascular Disease, Invasion/Metastasis, and Neurobiology research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty