Would you like to log in again?
Human Recombinant CRKL (from Baculovirus (Sf9 Insect Cells))
Supplier: Rockland Immunochemicals
009-001-R74S 009-001-R74
10800-912EA 313 USD InStock
10800-912 10800-914
Human Recombinant CRKL (from Baculovirus (Sf9 Insect Cells))
Proteins and Peptides
CRKL or v-crk sarcoma virus CT10 oncogene homolog (avian)-like is a protein kinase containing SH2 and SH3 (src homology) domains that activate the RAS and JUN kinase signaling pathways and transform fibroblasts in a RAS-dependent fashion (1). It is a substrate of the BCR-ABL tyrosine kinase which plays a role in fibroblast transformation by BCR-ABL, and may be oncogenic (2). CRKL is also required for normal cellular responses to Fgf8, including survival and migration, Erk activation, and target gene expression. CRKL Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Apoptosis/Autophagy, Cancer, Cell Cycle, Receptor Tyrosine Kinases, and WNT Signaling research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty