Would you like to log in again?
Human Recombinant CRYAA (from E. coli)
Supplier: Rockland Immunochemicals
009-001-R75 009-001-R75S
10800-930EA 313 USD InStock
10800-930 10800-928
Human Recombinant CRYAA (from E. coli)
Proteins and Peptides
CRYAA or crystallin alpha A is the major protein components of the vertebrate eye lens which is composed of 2 primary gene products--alpha-A and alpha-B. CRYAA can be induced by heat shock and are members of the small heat shock protein (sHSP also known as the HSP20) family (1). CRYAA plays an important role in the normal embryologic development of the anterior segment of the eye. In the Elo mouse, a 1-bp deletion in the gamma-E-crystallin gene causes autosomal dominant cataract and microphthalmia (2). CRYAA Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, Cancer, Metabolic Disorder, and Neurobiology research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty