Would you like to log in again?
Human Recombinant TAF1 (from Baculovirus (Sf9 Insect Cells))
Supplier: Rockland Immunochemicals
009-001-S28S 009-001-S28
75929-396EA 364 USD InStock
75929-396 75929-398
Human Recombinant TAF1 (from Baculovirus (Sf9 Insect Cells))
Proteins and Peptides
KAT4 is a DNA-binding protein complex required for RNA polymerase II-mediated transcription of many, if not all, protein-encoding genes in eukaryotic cells. KAT4 plays a key role in the initiation process, since it binds to the TATA element to form a complex that nucleates the assembly of the other components into a preinitiation complex and that may be stable through multiple rounds of transcription. KAT4 may be targeted to specific chromatin-bound promoters and may play a key role in chromatin recognition (1). KAT4 also serves to link the control of transcription to the cell cycle (2). KAT4 Protein is ideal for investigators involved in Signaling Proteins, Acetyl/Methyltransferases, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cell Cycle, ERK/MAPK Pathway, Inflammation, Invasion/Metastasis, Metabolic Disorder, Neurobiology, NfkB Pathway, and PKA/PKC Pathway research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty