Would you like to log in again?
Human Recombinant MDM2 (from E. coli)
Supplier: Rockland Immunochemicals
009-001-S45 009-001-S45S
75929-910EA 210 USD InStock
75929-910 75929-912
Human Recombinant MDM2 (from E. coli)
Proteins and Peptides
MDM2 is a nuclear phosphoprotein that binds and inhibits transactivation by p53, as part of an autoregulatory negative feedback loop (1). Overexpression of the MDM2 gene product can lead to excessive inactivation of p53 thereby diminishing its tumor suppressor function. The inactivation of p53 is mediated by the E3 ubiquitin ligase activity of MDM2 which targets p53 for proteasomal degradation. MDM2 also affects the cell cycle, apoptosis, and tumorigenesis through interactions with other proteins, including retinoblastoma 1 and ribosomal protein L5 (2). Amplification of MDM2 is frequently observed in human sarcomas and this is consistent with the hypothesis that MDM2 binds to p53 which then leads to escape from p53-regulated growth control. MDM2 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, AKT/PKB Pathway, Apoptosis/Autophagy, Cancer, Cell Cycle, and Cellular Stress research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty