Would you like to log in again?
Human Recombinant CAB39 (from Baculovirus (Sf9 Insect cells))
Supplier: Rockland Immunochemicals
009-001-S49 009-001-S49S
75929-976EA 364 USD InStock
75929-976 75929-978
Human Recombinant CAB39 (from Baculovirus (Sf9 Insect cells))
Proteins and Peptides
MO25α (mouse protein 25 alpha) is a 40-kDa protein that interacts with the STE20-related adaptor-alpha (STRADα) pseudo kinase to form a regulatory complex capable of stimulating the activity of the LKB1 tumor suppressor protein kinase (1). LKB1 plays a critical role in cell proliferation, polarity and energy metabolism. LKB1 is mutated in the inherited Peutz-Jeghers cancer syndrome (PJS). MO25α binds directly to a conserved Trp-Glu-Phe sequence at the STRADα C terminus, and markedly enhances the binding of STRADα to LKB1 thereby increasing LKB1 catalytic activity (2). MO25α Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cardiovascular Disease, and Metabolic Disorder research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty