Would you like to log in again?
Human recombinant Q53X90 (from E. coli)
Supplier: Rockland Immunochemicals
009-001-U66-1000 009-001-U66-0020 009-001-U66-0005 009-001-U66-0100
10800-962EA 326 USD InStock
10800-962 10801-032 10800-964 10801-030
Human recombinant Q53X90 (from E. coli)
Proteins and Peptides
B cell Attracting Chemokine-1 (BCA-1), also known as CXCL13, is expressed at high levels in lymphoid tissues such as the spleen, lymph nodes and Peyer’s patches. It has been shown to activate signaling through the receptor BLR1 (Burkitt’s lymphoma receptor 1) to chemoattract B cells. Recombinant human BCA-1 is a non-glycosylated protein, containing 87 amino acids, with a molecular weight of 10.3 kDa.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty