Would you like to log in again?
Human Recombinant CSF1 (from E. coli)
Supplier: Rockland Immunochemicals
009-001-V78-0002 009-001-V78-0100 009-001-V78-1000 009-001-V78-0010
75929-780EA 899 USD InStock
75929-780 75929-772 75929-784 75929-776
Human Recombinant CSF1 (from E. coli)
Proteins and Peptides
Macrophage Colony Stimulating Factor (M-CSF) is a hematopoietic growth factor produced by a wide variety of cells. M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells. Recombinant human M-CSF is a disulfide linked homodimer, containing two 159 amino acid chains, with a total molecular weight of 36.8 kDa.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty