Would you like to log in again?
Mouse Recombinant Kdm4b (from Baculovirus (Sf9 Insect Cells))
Supplier: Rockland Immunochemicals
010-001-S34S 010-001-S34
75929-418EA 364.21 USD InStock
75929-418 75929-420
Mouse Recombinant Kdm4b (from Baculovirus (Sf9 Insect Cells))
Proteins and Peptides
KDM4B or lysine (K)-specific demethylase 4B, which is also known as JMJD2B, contains a JmjN domain, a JmjC domain, a JD2H domain, 2 TUDOR domains, and a bipartite nuclear localization signal that overlaps the C-terminal part of the second TUDOR domain. KDM4B plays an essential role in human carcinogenesis through positive regulation of cyclin-dependent kinase 6 (1). KDM4B functions as a co-factor of estrogen receptor in breast cancer proliferation and mammary gland development (2). KDM4B is regulated by both ERa and HIF-1a, drives breast cancer cell proliferation in normoxia and hypoxia, and epigenetically regulates the expression of cell cycle genes such as CCND1, CCNA1, and WEE1. KDM4B Protein is ideal for investigators involved in Signaling Proteins, Deacetylase/Demethylase Proteins, Cancer, Cell Cycle, and Inflammation research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty