Would you like to log in again?
Mouse Recombinant MyI9 (from E. coli)
Supplier: Rockland Immunochemicals
010-001-S38 010-001-S38S
75929-564EA 364 USD InStock
75929-564 75929-566
Mouse Recombinant MyI9 (from E. coli)
Proteins and Peptides
LC20 is the myosin light chain that may regulate muscle contraction by modulating the ATPase activity of myosin heads (1). LC20 protein binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found for LC20 and the deduced 172-amino acid protein is highly conserved, with only 3 differences between the human and chicken proteins. Light chain phosphorylation causes the folded monomeric form of myosin to extend and assemble into filaments. This observation established the involvement of the LC20 in conformational transitions of smooth muscle myosin (2). LC20 Protein is ideal for investigators involved in Signaling Reagents, Protein Substrates, and Cardiovascular Disease research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty