Would you like to log in again?
Mouse Recombinant Csf3 (from E. coli)
Supplier: Rockland Immunochemicals
010-001-U93-0100 010-001-U93-0010 010-001-U93-0002 010-001-U93-1000
75928-194EA 137.15 USD InStock
75928-194 75928-188 75928-190 75928-192
Mouse Recombinant Csf3 (from E. coli)
Proteins and Peptides
Granulocyte Colony-Stimulating Factor, or G-CSF, is a growth factor that is considered the most potent inducer of terminal differentiation to granulocytes and macrophages of leukemic myeloid cell lines. The synthesis of G-CSF can be induced by bacterial endotoxins, TNF, IL-1 and GM-CSF. Prostaglandin E2 inhibits the synthesis of G-CSF, while in epithelial, endothelial, and fibroblastic cells secretion of G-CSF is induced by IL-17. Human and mouse G-CSF are cross-reactive. Recombinant mouse G-CSF is a non-glycosylated protein, containing 179 amino acids, with a molecular weight of 19 kDa.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty