Would you like to log in again?
Rat Recombinant Creb1 (from E. coli)
Supplier: Rockland Immunochemicals
012-001-R73S 012-001-R73
10800-900EA 368 USD InStock
10800-900 10800-902
Rat Recombinant Creb1 (from E. coli)
Proteins and Peptides
CREB1 is a member of the cAMP responsive element binding protein family of transcription factors that are critical mediators of gene expression in response to extracellular signals and are essential regulators of adaptive behavior and long-term memory formation (1). Activation of CREB1 by protein kinase A-mediated phosphorylation has been implicated in the survival of mammalian cells. Mice lacking CREB in the central nervous system during development show extensive apoptosis of postmitotic neurons (2). CREB binds as homo- and heterodimers to promoters containing CRE and activator protein 1 (AP-1) sites to alter target-gene expression). CREB1 Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, Apoptosis/Autophagy, Cardiovascular Disease, ERK/MAPK Pathway, Inflammation, Invasion/Metastasis, Metabolic Disorder, Neurobiology, NfkB Pathway, and PKA/PKC Pathway research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty