Would you like to log in again?
Rat Recombinant MIP-1 Alpha (CCL3)
Supplier: Rockland Immunochemicals
012-001-V80-0002 012-001-V80-0100 012-001-V80-0010 012-001-V80-1000
75929-790EA 667 USD InStock
75929-790 75929-792 75929-794 75929-788
Rat Recombinant MIP-1 Alpha (CCL3)
Proteins and Peptides
Macrophage Inflammatory Protein-1 alpha (MIP-1α), also known as CCL3, is produced by macrophages and is thought to induce inflammatory responses, including superoxide production by neutrophils. MIP-1α can exist as a naturally occurring heterodimer with MIP-1β and has been shown to have antiviral activity against HSV-1. Recombinant rat MIP-1α is a non-glycosylated protein, containing 69 amino acids, with a molecular weight of 7.8 kDa.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty