Would you like to log in again?
PERMAHAND® Silk Suture, No Needle, Ethicon
Supplier: Ethicon (Johnson & Johnson)
PERMA-HAND™
ETHA304H
95057-035BX 344.98 USD InStock
95057-035
PERMAHAND® Silk Suture, No Needle, Ethicon
Sutures
PERMAHAND® Silk Suture should be used by the person has experience with surgical procedures and techniques involving nonabsorbable sutures for wound closure. This has been followed a must to avoid the risk of wound dehiscence may vary with the site of application and the suture material used.
  • Do not resterilize and discard opened packages and unused sutures
  • Non-absorbable and unidirectional
Contraindicated in patients with known sensitivities or allergies to silk. Not recommended where permanent retention of tensile strength is required. Sutures are processed to remove the natural waxes and gums, dyed black, and coated with a special wax mixture.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty