Would you like to log in again?
SEMCO® Cartridges, Bergdahl Associates
Supplier: BERGDAHL ASSOCIATES INC. TE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
220315 220319
76240-682EA 10.65 USD InStock
76240-682 76254-248
SEMCO® Cartridges, Bergdahl Associates
Filters Filter Cartridges
The disposable plastic cartridges are made from LDPE and available in a variety of capacities.
All sizes are designed to accept ¹/₄" NPT fittings. Black cartridges are available for materials requiring protection from both visible and UV light.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty