Would you like to log in again?
Coiled Grounding Cord, SCS Division of Desco
Supplier: Desco Industries
2210
76238-056CS 466.68 USD InStock
76238-056
Coiled Grounding Cord, SCS Division of Desco
Cables
The wrist strap ground cords are a single bundle of tinsel conductors laced for great strength and reinforced with strong synthetic fibers.
  • Highly durable with segmented strain relief
  • Female snap socket, 4 mm
For use with single-wire wrist straps.
Certifications: Meets ANSI/ESD S20.20 tested per ANSI/ESD S1.1 and ESD TR53.
Delivery information: Includes alligator clip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty