Would you like to log in again?
Economy Heel Grounder, Desco Industries
Supplier: Desco Industries
2044
76240-018BX 1028.6 USD InStock
76240-018
Economy Heel Grounder, Desco Industries
Grounders Heel and Toe Grounders
Economy heel grounder assembly is a cup-style heel grounder with a high visibility strap that helps facility supervisors effectively manage their static control program.
  • Dependable and cost-efficient
  • Used to ground operator to ESD Flooring
  • Adjustable sizing material expands to fit most shoe sizes comfortably
Allows supervisors to see at a single glance whether employees are properly wearing their heel grounding assemblies, and to help ensure their company is protected against static damage.
Certifications: Meets the requirements of ANSI/ESD S20.20.
Caution: This product is not recommended for use on equipment with operating voltage exceeding 250 V.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty