Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Wescorp ESD Conductive Grid Tape, Desco

Supplier: Desco Industries
81251 81250 81252 81253
76243-844EA 34.42 USD InStock
76243-844 76242-024 76244-484 76243-296
Wescorp ESD Conductive Grid Tape, Desco
Tapes
Thes tapes with low charging transparent cellulose film with high-tack rubber adhesive.

  • Conductive shielding layer (1×104 to <1×106 ohms) marked with ESD symbols (MIL STD 1285 and ESDA)
  • Surface resistance 1×108 to <1×1011 ohms
  • Temperature range 10 to 71 °C (14 - 140 °F)
  • Meets MIL-B-81705 for exposure to 100 °C for 10 minutes max

Wescorp tape generates less than 50 volts when unwound from roll (at 73 °F, 45% RH average) making it safe to use around most ESD sensitive items.

Used in creating a Faraday cage to provide static shielding of an ESD sensitive item.

All tapes will generate some amount of triboelectric charge when unwound from roll. If voltage less than 50 volts required, a grounded tape dispenser or ionization is recommended when unrolling the tape.

Certifications: Tested per ANSI/ESD S11.11 to meet dissipative limits of packaging standard ANSI/ESD S541.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty