Would you like to log in again?
MAPK3 Blocking Peptide
Supplier: Rockland Immunochemicals
000-001-GP9
76222-528EA 154 USD InStock
76222-528
MAPK3 Blocking Peptide
Proteins and Peptides
ERK1 C-Term Blocking Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Mitogen activiated protein kinase 3, also known as MAPK3, ERK, or ERK1, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. ERK1 and ERK2 are activated by MEK1 and MEK2 in the B-raf signaling pathway resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Human ERK1 and ERK2 are 84% identical in sequence and share common functionality in cells. ERK1 is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
ERK1 C-term blocking peptide is suitable as a control for polyclonal or monoclonal anti-ERK1 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty