Would you like to log in again?
HCP Control
Supplier: Rockland Immunochemicals
000-001-J08
RL000001J08EA 162 USD InStock
RL000001J08
HCP Control
Proteins and Peptides
E.coli HCP Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
  • 1-D electrophoresis
  • 2D electrophoresis
Host Cell Protein Lysate was prepared by solubilizing mock induced E.coli cells. Host Cell Proteins are process-related impurities derived from a host cell expression system that may be present in trace amounts in a final drug substance. HCP Antibodies are used to detect impurities in biologically-based therapeutics. Detecting impurities often unseen by 1-D electrophoresis, a 2D electrophoresis western blot analysis delivers more complete insight into the immunocoverage of protein components of the host cell lysate.
E. coli Host Cell Protein Control Proteins detects over 300 E.coli proteins and is suitable for western blotting and ELISA.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty