Would you like to log in again?
CEFT panel 32 peptides
Supplier: Rockland Immunochemicals
000-001-L24
RL000-001-L24EA 2638 USD InStock
RL000-001-L24
CEFT panel 32 peptides
Proteins and Peptides
Pool of 32 HLA class I restricted T cell epitopes from human cytomegalovirus, Epstein-Barr virus and influenza virus.
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Pool of 32 HLA class I restricted T cell epitopes from human cytomegalovirus, Epstein-Barr virus and influenza virus. Stimulates the release of IFN-gamma from CD8+ T cells in individuals with certain highly frequent HLA types (HLA: A1, A2, A3, A11, A24, A68, B7, B8, B27, B35, B44). Chosen epitopes are presented by the most common Caucasian HLA types, whose cumulative frequencies represent >100% of Caucasian individuals. Useful as a peptide specific positive control in ELISPOT, CTL and intracellular cytokine assays using human PBMC.
T-Cell Assay Panel Control Peptides are suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty