Would you like to log in again?
MOG 35-55-OH
Supplier: Rockland Immunochemicals
000-001-M41
RL000-001-M41EA 148 USD InStock
RL000-001-M41
MOG 35-55-OH
Proteins and Peptides
Myelin oligodendrocyte glycoprotein (MOG) Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Myelin oligodendrocyte glycoprotein (MOG) is a minor constituent of the CNS myelin synthesized by oligodendrocytes and Schwann cells and is thought to mediate homophilic cell-cell adhesion among other roles. MOG, Myelin basic protein (MBP), and proteolipid proteins (PLP) are potential primary encephalitogenic target antigens in multiple sclerosis.
Myelin oligodendrocyte glycoprotein (MOG) Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty