Would you like to log in again?
HIV-1 tat, 48-60 Peptide
Supplier: Rockland Immunochemicals
000-001-M49
RL000-001-M49EA 112 USD InStock
RL000-001-M49
HIV-1 tat, 48-60 Peptide
Proteins and Peptides
HIV-1 tat protein Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Translocation through the plasma membrane has been shown to be a major limiting step for the delivery of various macromolecules to the cytoplasm and other intracellular compartments (e.g., mitochondria, nucleus). Numerous studies have confirmed that specific peptide sequences known as cell penetrating peptides (CPP) derived from proteins able to cross the plasma membrane, can be added to various cargo and delivered across cell membranes. The cargo molecules that have been successfully transported into cells includes oligonucleotides, peptides, peptide nucleic acids, proteins and nanoparticles. One of these translocating peptides was derived from the HIV-1 Tat protein, specifically located within the first exon of the HIV tat protein. The specific HIV tat sequence is highly basic (cationic) and is readily added to peptides either as a preformed peptide with a site for direct conjugation to other molecules (typically a cysteine). Addition of the tat-cargo complex (5-50 uM concentration) to cells for 30-60 minutes results in the transfer of the tat-cargo complex to intracellular locations in a rapid, dose-dependent manner. The addition of nuclear or mitochondrial localization sequences has been shown to specifically direct the cargo to the nucleus or mitochondria respectively.
HIV-1 tat, 48-60 Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty