Would you like to log in again?
Human Recombinant LEP
Supplier: Rockland Immunochemicals
009-001-320
RL009-001-320EA 326 USD InStock
RL009-001-320
Human Recombinant LEP
Proteins and Peptides
Leptin Human Recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Leptin inhibits food intake and stimulates energy expenditure. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe hereditary obesity in rodents and humans is caused by defects in leptin production. In addition to its critical role in the physiologic regulation of body weight leptin has a variety of other physiologic and pathologic functions resembling those of cytokines. These functions include the regulation of hematopoiesis, angiogenesis, wound healing, inflammation, and immune responses. Leptin Human Recombinant produced in E. coli is a single, non-glycosylated, polypeptide chain containing 147 amino acids and having a molecular mass of 16 kDa.
Leptin is suitable as a control for polyclonal or monoclonal anti-Leptin in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty