Would you like to log in again?
HSPD1
Supplier: Rockland Immunochemicals
009-001-F62S 009-001-F62
75928-766EA 714 USD InStock
75928-766 75928-764
HSPD1
Proteins and Peptides
HSP60 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
HSP60 is a chaperonin protein, required for ATP-dependent folding of precursor polypeptides and complex assembly and prevents aggregation and mediates protein refolding after heat shock. Mutations and variations in the HSP 60 gene product may be implicated in genetic diseases (1). HSP60 has been strongly implicated as an example of molecular mimicry in the pathogenicity of autoimmune diseases in T cell-mediated protection. Human HSP60 was found to activate the complement system in normal human serum in a dose-dependent manner. Since complement activation plays an important role in the development of atherosclerosis, the levels of complement-activating anti-HSP60 antibodies are elevated in atherosclerosis-related diseases. (2) HSP60 Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, Cancer, Cardiovascular Disease, Cellular Stress, and Inflammation research.
HSP60 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. HSP60 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~85kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty