Would you like to log in again?
HSP90AB1
Supplier: Rockland Immunochemicals
009-001-F71 009-001-F71S
75934-606EA 714 USD InStock
75934-606 75934-604
HSP90AB1
Proteins and Peptides
HSP90 beta control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
HSP90β is a member of the HSP90 family of proteins which are important molecular chaperones involved in signal transduction, cell cycle control, stress management, and folding, degradation, and transport of proteins (1). HSP90 proteins have been found in a variety of organisms suggesting that they are ancient and conserved. HSP90 binds to client proteins (such as steroid receptors, AKT, Bcr-Abl, Apaf-1, survivin, cyclin dependent kinases) and acts as a molecular chaperone. Failure of Hsp90 chaperone activity leads to misfolding of client proteins, which leads to ubiquitination and proteasome degradation, and thus deregulation of cellular homeostasis (2). HSP90β Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, AKT/PKB Pathway, Apoptosis/Autophagy, Cancer, and Cellular Stress research.
HSP90β Protein is stored in 50mM NaPhosphate pH7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.2mM DTT, 25% glycerol. HSP90β Protein is suitable for use in Western Blot and ATPase Assay. Expect a band approximately ~91kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty