Would you like to log in again?
SOD2
Supplier: Rockland Immunochemicals
009-001-G14S 009-001-G14
75932-092EA 364 USD InStock
75932-092 75932-094
SOD2
Proteins and Peptides
SOD2 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SOD2 (superoxide dismutase 2) is a member of the iron/manganese superoxide dismutase family. SOD2 binds to the superoxide byproducts of the mitochondrial electron transport chain and converts them to hydrogen peroxide and diatomic oxygen. Failure of SOD2 to remove the superoxide byproducts leads to an increase in mitochondrial reactive oxygen species resulting in biochemical aberrations with features reminiscent of mitochondrial myopathy, Friedreich ataxia, and HMGCL deficiency (1). Mutations in SOD2 have been associated with idiopathic cardiomyopathy (IDC), premature aging, sporadic motor neuron disease, and cancer (2). SOD2 Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cellular Stress, Inflammation, Neurobiology, and p38 Pathway research.
SOD2 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. SOD2 Protein is suitable for use in Western Blot. Expect a band approximately ~51kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty