Would you like to log in again?
Human Recombinant IL-22
Supplier: Rockland Immunochemicals
009-001-J33-0002 009-001-J33-0010 009-001-J33-1000 009-001-J33-0100
75928-798EA 326 USD InStock
75928-798 75928-796 75928-802 75928-800
Human Recombinant IL-22
Proteins and Peptides
rHuman IL-22 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-22 (IL-22), also called IL-TIF, is an IL-10 family member, produced by activated dendritic cells and T lymphocytes. IL-22 signals via the IL-22R and IL-10Rbeta receptors to act as a potent mediator of the cellular inflammatory response. Recombinant human IL-22 is a non-disulfide-linked homodimer, containing two 146 amino acids, with a total molecular weight of 33.6 kDa.
Interleukin-22 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-22 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty