Would you like to log in again?
HO2 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S04 009-001-S04S
75928-508EA 364 USD InStock
75928-508 75928-510
HO2 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
HO2 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
HO2, or heme oxygenase 2, belongs to the heme oxygenase family that are essential enzyme in heme catabolism that cleave heme to form biliverdin which is subsequently converted to bilirubin by biliverdin reductase (1). HO2 is part of the calcium-sensitive potassium channel complex and enhances channel activity in normoxia (2). Knockdown of HO2 expression reduces channel activity, an effect that is reversed by carbon monoxide. Inhibition of calcium-sensitive potassium channels by hypoxia is dependent on HO2 expression and is augmented by HO2 stimulation. HO2 Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, Cardiovascular Disease, Cellular Stress, and Neurobiology research.
HO2 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. HO2 Protein is suitable for use in Western Blot. Expect a band approximately ~62kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty