Would you like to log in again?
INCENP
Supplier: Rockland Immunochemicals
009-001-S21 009-001-S21S
75928-938EA 714 USD InStock
75928-938 75928-936
INCENP
Proteins and Peptides
INCENP control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
INCENP or inner centromere protein antigens 135/155kDa is a constitutively binding centromere proteins and 'passenger,' or transiently interacting, proteins. The overexpression of a truncation mutant of chicken INCENP and a chimeric CENPB:INCENP protein in mammalian cell culture disrupted both prometaphase chromosome alignment and completion of cytokinesis (1). INCENP forms a complex with the evolutionarily conserved family of Aurora B kinases which helps coordinate chromosome segregation, spindle behavior, and cytokinesis during mitosis (2). INCEP Protein is ideal for investigators involved in Signaling Proteins, Microtubule/Actin Associated Proteins, Cell Cycle, and Invasion/Metastasis research.
INCENP Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. INCENP Protein is suitable for use in Western Blot. Expect a band approximately ~19kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty