Would you like to log in again?
CREBBP
Supplier: Rockland Immunochemicals
009-001-S26S 009-001-S26
75929-388EA 364 USD InStock
75929-388 75929-390
CREBBP
Proteins and Peptides
KAT3A (CREBBP) control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
KAT3A has intrinsic histone acetyltransferase activity that acts as a scaffold to stabilize additional protein interactions with the transcription complex and acetylates both histone and non-histone proteins. KAT3A (CREBBP) expressed as a nuclear protein that binds to cAMP-response element binding protein (CREB) and involved in the transcriptional coactivation of many different transcription factors. KAT3A plays a main role in embryonic development, growth control, and homeostasis by coupling chromatin remodeling to transcription factor recognition (1). KAT3A also plays a critical role in the transmission of inductive signals from cell surface receptors to the transcriptional apparatus (2). KAT3A Protein is ideal for investigators involved in Signaling Proteins, Acetyl/Methyltransferase Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cell Cycle, ERK/MAPK Pathway, Inflammation, Invasion/Metastasis, Metabolic Disorder, Neurobiology, NfkB Pathway, and PKA/PKC Pathway research.
KAT3A Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. KAT3A Protein is suitable for use in Western Blot. Expect a band approximately ~65kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty