Would you like to log in again?
ELP3
Supplier: Rockland Immunochemicals
009-001-S30 009-001-S30S
75929-408EA 714 USD InStock
75929-408 75929-406
ELP3
Proteins and Peptides
KAT9 (ELP3) control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
KAT9 (also known as ELP3) is the catalytic subunit of the histone acetyltransferase (HAT) elongator complex, which contributes to transcript elongation and also regulates the maturation of projection neurons. The knockdown of KAT9 by antisense morpholinos in zebrafish embryos resulted in dose-dependent shortening and abnormal branching of motor neurons with no concomitant morphologic abnormalities (1). KAT9 knockdown also impairs paternal DNA demethylation as indicated by reporter binding, immunostaining, and bisulfite sequencing (2). KAT9 Protein is ideal for investigators involved in Signaling Proteins, Acetyl/Methyltransferase Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cell Cycle, ERK/MAPK Pathway, Inflammation, Invasion/Metastasis, Metabolic Disorder, Neurobiology, NfkB Pathway, and PKA/PKC Pathway research.
KAT9 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. KAT9 Protein is suitable for use in Western Blot. Expect a band approximately ~86kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty