Would you like to log in again?
KDM2A
Supplier: Rockland Immunochemicals
009-001-S32S 009-001-S32
75933-794EA 364 USD InStock
75933-794 75933-796
KDM2A
Proteins and Peptides
KDM2A control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
KDM2A or lysine (K)-specific demethylase 2A is a member of the F-box protein family that are components of the modular E3 ubiquitin protein ligases called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination (1). KDM2A belongs to the Fbls class and, in addition to the presence of an F-box, contains at least six highly degenerated leucine-rich repeats which play a role in epigenetic silencing. The demethylase activity of the JmjC domain-containing proteins is conserved from yeast to human (2). KDM2A Protein is ideal for investigators involved in Signaling Proteins, Deacetylase/Demethylase Proteins, Cancer, Cell Cycle, and Inflammation research.
KDM2A is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. KDM2A Protein is suitable for use in Western Blot. Expect a band approximately ~115kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty