Would you like to log in again?
p63 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S68S 009-001-S68
75930-724EA 714 USD InStock
75930-724 75930-722
p63 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
p63 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
p63 is the primordial member of the p53 family that acts in a conserved process of monitoring the integrity of the female germline, whereas the functions of p53 are restricted to vertebrate somatic cells for tumor suppression. p63 protein plays an important role in the development and maintenance of stratified epithelial tissues (1). p63 is critical for maintaining the progenitor-cell populations that are necessary to sustain epithelial development and morphogenesis (2). Mutations in p63 are associated with ectodermal dysplasia, cleft lip/palate syndrome 3 (EEC3) and split-hand/foot malformation 4 (SHFM4). p63 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cell Cycle, Cellular Stress, Inflammation, JNK/SAPK Pathway, Metabolic Disorder, Neurobiology, and p38 Pathway research.
p63 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. p63 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~92kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty