Would you like to log in again?
KAT2B
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
009-001-S81S 009-001-S81
75930-744EA 714 USD InStock
75930-744 75930-742
KAT2B
Proteins and Peptides
PCAF control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PCAF is a protein that shows homology to the yeast GCN5 and associates with p300/CBP. PCAF has in vitro and in vivo binding activity with CBP and p300, and competes with E1A for binding sites in p300/CBP (1). PCAF encodes a predicted 832-amino acid protein that showed by RNA blotting to be expressed in all tissues, most strongly in heart and skeletal muscle. PCAF has histone acetyl transferase activity with core histones and nucleosome core particles, indicating that this protein plays a direct role in transcriptional regulation. PCAF is required for MyoD activity and muscle differentiation. PCAF directly acetylated MyoD which then displays an increased affinity for its DNA target (2). PCAF Protein is ideal for investigators involved in Signaling Proteins, Acetyl/Methyltransferase Proteins, Cancer, Cell Cycle, and NfkB Pathway research.
PCAF Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PCAF Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~69kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty